Frocks, PP
Green, XXL
Catalog # HOPE40.301V/05
Supplier: Medicom
Aller à :
Product Details & Documents
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
- With collar and long sleeves
- Elasticated cuffs
- Snap buttons
- Without pockets
Weight: 40 g/m²
Spécifications
- Clothing Size:XXL
- Colour:Green
- Cat. No.:HOPE40.301V/05
- Garment application:General Purpose|Industrial
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Weight:40 g/m²