ContactUsLinkComponent
A potent, dual PI3K/mTOR inhibitor (IC50 values are 0.4 and 5.4 nM for PI3K and PI3K, respectively, and 1.6 nM for mTOR) 1 * 10 mg
Catalog # CAYM14567-10
Supplier:  Cayman Chemical
A potent, dual PI3K/mTOR inhibitor (IC50 values are 0.4 and 5.4 nM for PI3K and PI3K, respectively, and 1.6 nM for mTOR) 1 * 10 mg
Catalog # CAYM14567-10
Supplier:  Cayman Chemical
Frequently Bought Together