ContactUsLinkComponent
An inhibitor of UCH-L1 activity (IC50 = 0.88 Ki = 0.4 M) that demonstrates selectivity for UCH-L1 compared to UCH-L3 (IC50 = 25 M) 1 * 5 mg
Catalog # CAYM15396-5
Supplier:  Cayman Chemical
An inhibitor of UCH-L1 activity (IC50 = 0.88 Ki = 0.4 M) that demonstrates selectivity for UCH-L1 compared to UCH-L3 (IC50 = 25 M) 1 * 5 mg
Catalog # CAYM15396-5
Supplier:  Cayman Chemical
Frequently Bought Together