Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin NF

Vanillin NF

Poly pail, 2,5 kg
Catalog # SPCMVA130-2.5KG
Irritant
Supplier:   Spectrum Chemical
Vanillin NF
Catalog # SPCMVA130-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  121-33-5
Vanillin NF
Vanillin NF

Poly pail, 2,5 kg

CAS Number:  121-33-5
Price
836,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specification Test Results

  • Assay:
    97% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    2,5 kg

Specifications