Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Proline USP

L(-)-Proline USP

Drum, 5 kg
Catalog # SPCMP1434-5KG
Supplier:   Spectrum Chemical
L(-)-Proline USP
Catalog # SPCMP1434-5KG
Supplier:  Spectrum Chemical
CAS Number:  147-85-3
L(-)-Proline USP

Drum, 5 kg

CAS Number:  147-85-3
Price
4 370,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
  • CAS Number:147-85-3
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00064318
  • Molecular Weight:115.13 g/mol
  • Merck Index:13,07871
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:220…222 °C
  • Density:1.35 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:205-702-2
Specifications

Specifications

  • Pack type:
    Drum
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.