Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic, powder USP

Bismuth(III) nitrate basic, powder USP

12 kg
Catalog # SPCMBI150-12KG
Oxidizing
Irritant
Supplier:   Spectrum Chemical
Bismuth(III) nitrate basic, powder USP
Catalog # SPCMBI150-12KG
Supplier:  Spectrum Chemical
CAS Number:  1304-85-4
Bismuth(III) nitrate basic, powder USP

12 kg

CAS Number:  1304-85-4
Price
3 800,00 €
Log in for availability
1 * 12 kg
Product Details & Documents

Form: Powder

Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Density:4.928 g/cm³ (20 °C)
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Specifications

Specification Test Results

  • Assay:
    79% Min.

Specifications

  • Pk:
    12 kg

Specifications