Cholesterol NF
Poly pail, 2,5 kg
Catalog # SPCMCH135-2.5KG
Supplier: Spectrum Chemical
Aller à :
Product Details & Documents
BSE/TSE statement (available upon request) Allergen statement (available upon request)
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
: LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
Synonymes: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- Numéro CAS:57-88-5
- Formule:C₂₇H₄₆O
- Numéro MDL:MFCD00003646
- Poids moléculaire:386.66 g/mol
- Index Merck:13,02221
- Point d'ébullition:360 °C (1013 hPa)
- Point de fusion:147…150 °C
- Densité:1.07 g/cm³ (20 °C)
- Température de stockage:Ambient
- EINECS:200-353-2
Spécifications
Résultats des tests de spécification
- ASSAY (DRIED BASIS):95.0 - 102.0 %
- MELTING RANGE:147° - 150°C
- SPECIFIC ROTATION:-34° to -38°
- SOLUBILITY IN ALCOHOL:No deposit or turbidity forms
- ACIDITY:max 0.3 ml
- LOSS ON DRYING:max 0.3%
- RESIDUE ON IGNITION:max 0.1%
- β-CHOLESTANOL (5a-CHOLESTAN-3β-ol,DIHYDROCHOLESTEROL):max 0.6 %
- DESMOSTEROL:max 4%
- LATHOSTEROL:max 2%
- 24-DEHYDROLATHOSTEROL (5a-CHOLESTA-7,24 DIEN--3β-ol ):max 0.2 %
- 4-METHYLCHOLEST-5-EN-3β-OL:max 0.5 %
- TOTAL IMPURITIES INCLUDING RELATED STEROLS:max 5%
- ELEMENTAL IMPURITIES:AS REPORTED
- IDENTIFICATION (A):SPECTRUM MATCHES REFERENCE
- IDENTIFICATION (B):-34° to -38°
- IDENTIFICATION (C):147 - 150 °C
Spécifications
- Pack type:Poly pail
- Pk:2,5 kg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



