ContactUsLinkComponent
SONOREX SUPER RK 1028 C, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 500X300X300 MM 45 L, S/S AISI 316 TI TIMER 1-15 MIN AND CON TINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 2000 W, HF-OUTPUT: 500 WEFF. 1 * 1 items
Catalog # BAND661-GB
SONOREX SUPER RK 1028 C, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 500X300X300 MM 45 L, S/S AISI 316 TI TIMER 1-15 MIN AND CON TINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 2000 W, HF-OUTPUT: 500 WEFF. 1 * 1 items
Catalog # BAND661-GB
Frequently Bought Together