Tissue processing/embedding cassettes, Unisette™, M405
Blue
Catalog # SIMPM405-6
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M405
Catalog # SIMPM405-6
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These cassettes are moulded from a special high density polymer and keep specimens safely submerged in liquid.
- Resistant to the chemical action of most histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- One-piece integral lid eliminates the need for separate steel lids
- Snap latch and hinge-lock design prevent early separation of base and lid
Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss.
Ordering information: Available in non-cytotoxic, non-metallic colors.
Packaging: Available in 25 stacks of 40 cassettes or 3 dispenser boxes of 500 cassettes.
Documents
Specifications
- Colour:Blue
- Seq:05
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



