Orders LinkContactUsLinkComponent
Separating funnel, pear shaped
Separating funnel, pear shaped
250 ml, 19/26 NS
Catalog # SMIT8S152/250
Supplier:   SMITH SCIENTIFIC
Separating funnel, pear shaped
Catalog # SMIT8S152/250
Supplier:  SMITH SCIENTIFIC
Separating funnel, pear shaped
250 ml, 19/26 NS
Price
132,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3, with plastic stopper.
  • Stopcock with glass key
  • Very good temperature resistance
  • Very good chemical resistance
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    250 ml