Multiscreen® High Volume Filter Plates
MultiScreen® High volume filter plates, PP, Opaque, Glass fiber membrane, 1,2 µm, 96, -
Catalog # MILFMVFCN1225
Supplier: Merck
Jump to:
Product Details & Documents
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
- Sample prep
- Protein kinase/phosphatase assays
- Receptor/ligand binding assays
Specifications
- Membrane:Glass fiber membrane
- No. of wells:96
- Pore size:1,2 µm
- Sterile:-
- Colour:Opaque
- Description:MultiScreen® High volume filter plates
- Material:PP
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



