Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test tubes

Test tubes

5 ml, 48,9 mm, 15,5 mm, flat bottom, graduated, 13,7 mm
Catalog # DELT401100
Supplier:   DELTALAB
Test tubes
Catalog # DELT401100
Supplier:  DELTALAB
Test tubes
Test tubes

5 ml, 48,9 mm, 15,5 mm, flat bottom, graduated, 13,7 mm

Price
264,00 €
Log in for availability
1 * 5.000 ST
Product Details & Documents

Polypropylene (PP), ultra clear.

  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable

Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.

Specifications
  • Ext.Ø:
    15,5 mm
  • Ø int.:
    13,7 mm
  • Version:
    flat bottom, graduated
  • Capacity:
    5 ml
  • Length:
    48,9 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.