Orders LinkContactUsLinkComponent
Face shield, Turboshield™
Face shield, Turboshield™
Chemical and impact resistant, PC, 240x 230 mm
Catalog # DALL1031743
Supplier:   HONEYWELL SAFETY
Face shield, Turboshield™
Catalog # DALL1031743
Supplier:  HONEYWELL SAFETY
Face shield, Turboshield™
Chemical and impact resistant, PC, 240x 230 mm
Price
22,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Turboshield™ is a great value faceshield that offers easy visor exchange, is more comfortable to wear and offers better protection.
  • Simple push‐button release system allows for easy visor exchange in seconds
  • Breathable, removable, washable headband
  • Toric lens provides excellent optics, increased field of view and added chin coverage without bulky attachments
  • Ergonomic, easy‐to‐use adjustment knobs
Applications include grinding, welding, chemical, construction, food services, manufacturing, medical, oil and gas, pharmaceutical, steel and metals.
Certifications: AS/NZS 1337.1:2010
Specifications
  • Type:
    Chemical and impact resistant
  • L×W:
    240x 230 mm
  • Material:
    PC
  • Seq:
    01