Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Phenylethanol USP

2-Phenylethanol USP

Glass, amber, 1 kg
Catalog # SPCMP1058-1KG
Toxic
Supplier:   Spectrum Chemical
2-Phenylethanol USP
Catalog # SPCMP1058-1KG
Supplier:  Spectrum Chemical
CAS Number:  60-12-8
2-Phenylethanol USP

Glass, amber, 1 kg

CAS Number:  60-12-8
Price
459,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Merck Index:12,07370
  • Boiling Pt:432 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Density:1.02 g/cm³ (20 °C)
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • EINECS:200-456-2
  • UN:2810
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    1 kg

Specifications