ContactUsLinkComponent
Microscope slides Unimark, 76x26mm, thickness 1mm, white glass, edges ground at 90°, white labelling area, wih 1 circle of 15mm i.d., w/out orientation mark, in boxes of 50 pcs., 2500 pcs./carton 1 * 2.500 items
Catalog # MARI0704062
Supplier:  MARIENFELD
Microscope slides Unimark, 76x26mm, thickness 1mm, white glass, edges ground at 90°, white labelling area, wih 1 circle of 15mm i.d., w/out orientation mark, in boxes of 50 pcs., 2500 pcs./carton 1 * 2.500 items
Catalog # MARI0704062
Supplier:  MARIENFELD
Frequently Bought Together