Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standards, common and transition elements, VHG

Multi-element standards, common and transition elements, VHG

Common and transition element standards, 500 ml, 500 mL
Catalog # PROOVHG-SM75B-500
Multi-element standards, common and transition elements, VHG
Catalog # PROOVHG-SM75B-500
CAS Number:  
Multi-element standards, common and transition elements, VHG

Common and transition element standards, 500 ml, 500 mL

CAS Number:  
Price
1 400,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).

Multi elements standards for Ag, Al, As, Ba, Be, Bi, Cd, Cr, Co, Cu, Fe, Li, Mn, Mo, Ni, Pb, Sb, Se, Sr, Tl, V, Zn at 100 µg/ml and Ca, K, Mg, Na at 1000 µg/ml in 0,2% HF.

Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Common and transition element standards
  • Pk Info:
    500 ml

Specifications