Multi-element standards, common elements mix 2, VHG
Product Details & Documents
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES), and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V and Zn.
Certificeringen: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Specifications
Specifications
- Concentration:100 mg/L
- Matrix:5% HCl/tr HF
- Pk:100 mL
- Description:Common elements mix 2
- Pk Info:100 ml
Specifications
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



