ContactUsLinkComponent
Rotavapor R-300 no bath, no joint., A, WB, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300200A011
Fournisseur:  BUCHI
Rotavapor R-300 no bath, no joint., A, WB, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300200A011
Fournisseur:  BUCHI
Frequently Bought Together