Orders LinkContactUsLinkComponent
Tubes and systems for active sampling
Tubes and systems for active sampling
Activated charcoal tubes, BIA
Catalog # DRAE6400743
Supplier:   Dräger
Tubes and systems for active sampling
Catalog # DRAE6400743
Supplier:  Dräger
Tubes and systems for active sampling
Tubes and systems for active sampling
Activated charcoal tubes, BIA
Price
320,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
  • High precision guaranteed
  • Very reliable in use
  • Improved colour depth and contrast using an optimised production method
Specifications
  • Type:
    BIA
  • Description:
    Activated charcoal tubes
  • Seq:
    03