Microscope slides, Corning®
Plain slides, Colourless, Glass, 75×50 mm
Catalog # CORN2947-75X50_72
Supplier: Corning
Aller à :
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
- Clarity reinforced water-white glass
- Frosted slides are sandblasted, easy to write on and highly legible
- Each slide is inspected for chips and rough edge before being shipped
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Colour:Colourless
- Description:Plain slides
- L×W:75×50 mm
- Material:Glass
- Seq:06