ContactUsLinkComponent
Volumetric flasks with one circular mark, 10 ml, +/- 0,04 ml, cl. A conformity certified with batch certificate, borosilicate glass 3.3, clear glass, blue graduated, with ground joint and polystopper, NS 7/16, 10pcs./carton, MARIENFELD SUPERIOR GTIN: 1 * 10 items
Catalog # MARI2332182
Supplier:  MARIENFELD
Volumetric flasks with one circular mark, 10 ml, +/- 0,04 ml, cl. A conformity certified with batch certificate, borosilicate glass 3.3, clear glass, blue graduated, with ground joint and polystopper, NS 7/16, 10pcs./carton, MARIENFELD SUPERIOR GTIN: 1 * 10 items
Catalog # MARI2332182
Supplier:  MARIENFELD
Frequently Bought Together