Orders LinkContactUsLinkComponent
Software for Evolution™ UV/VIS Spectrophotometers
Software for Evolution™ UV/VIS Spectrophotometers
Visionlite colorcalc, to measure color, includes software to perform color calculations from full spectrum scan of Evolution spectrophotometers
N° de catalogue SPET869-175600
Fournisseur:   Thermo Fisher Scientific
Software for Evolution™ UV/VIS Spectrophotometers
N° de catalogue SPET869-175600
Software for Evolution™ UV/VIS Spectrophotometers
Software for Evolution™ UV/VIS Spectrophotometers
Visionlite colorcalc, to measure color, includes software to perform color calculations from full spectrum scan of Evolution spectrophotometers
Prix
4 390,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Visionlite colorcalc, to measure color, includes software to perform color calculations from full spectrum scan of Evolution spectrophotometers
  • Seq:
    03