Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Straight, 150 mm
Catalog # BOCH1611
Supplier:   BOCHEM
Forceps, precision
Catalog # BOCH1611
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Straight, 150 mm
Price
13,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production and more.
  • Extremely sharp, flat
  • Non serrated points
Specifications
  • Type:
    Straight
  • Length:
    150 mm