ContactUsLinkComponent
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperature sensor: Pt100, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric e 1 * 1 items
Catalog # HAMI242428-1121
Supplier:  HAMILTON BONADUZ
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperature sensor: Pt100, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric e 1 * 1 items
Catalog # HAMI242428-1121
Supplier:  HAMILTON BONADUZ
Frequently Bought Together