Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 30×20 ml, 2,00, 30
Catalog # METT30111134
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT30111134
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 30×20 ml, 2,00, 30

CAS Number:  
Price
79,80 €
Log in for availability
1 * 30 ST
Specifications

Specifications

  • Pk:
    30
  • Description:
    Technical buffer solution, 30×20 ml
  • pH value:
    2,00

Specifications