pH Buffer Solution, VWR Chemicals BDH®
10,0, 500 ml
Catalog # BDHBBDH50720-500A
Supplier: BDH US PRODUCED
pH Buffer Solution, VWR Chemicals BDH®
Catalog # BDHBBDH50720-500A
Supplier: BDH US PRODUCED
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:500 ml
- pH value:10,0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



