Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffer Solution, VWR Chemicals BDH®

pH Buffer Solution, VWR Chemicals BDH®

7,0, 500 ml
Catalog # BDHBBDH50460-500A
Supplier:   BDH US PRODUCED
pH Buffer Solution, VWR Chemicals BDH®
Catalog # BDHBBDH50460-500A
Supplier:  BDH US PRODUCED
CAS Number:  
pH Buffer Solution, VWR Chemicals BDH®

7,0, 500 ml

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 ml
  • pH value:
    7,0

Specifications