Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Vanadium(V) oxide 99+%

Glass bottle, 25 g
Catalog # ACRO450160250
Corrosive
Irritant
Health
Environment
Vanadium(V) oxide 99+%
Catalog # ACRO450160250
CAS Number:  1314-62-1
Vanadium(V) oxide 99+%
Vanadium(V) oxide 99+%

Glass bottle, 25 g

CAS Number:  1314-62-1
Price
20,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Boiling Pt:1750 °C (760 mmHg)
  • Melting Pt:690 °C
  • Density:3.35 g/cm³
  • Storage Temperature:Ambient
  • UN:2862
  • ADR:6.1,III

 

Specifications

Specification Test Results

  • Appearance:
    Dark yellow Powder
  • Assay:
    ≥99.0 %
  • Particle size:
    ≤1.2 mm
  • Chromium (Cr):
    ≤0.1 %
  • Iron (Fe):
    ≤0.1 %
  • Phosphorus (P):
    ≤0.04 %
  • Potassium (K):
    ≤0.4 %
  • Silicon (Si):
    ≤0.06 %
  • Sodium (Na):
    ≤0.3 %
  • Sulfur (S):
    ≤0.07 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications