Lead(II) acetate basic ACS
Plastic bottle, 5 kg
Supplier: Thermo Fisher Scientific
Lead(II) acetate basic ACS
Catalog # ACRO453220050
Supplier: Thermo Fisher Scientific
CAS Number: 51404-69-4
Jump to:
Product Details & Documents
Synonyms: Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
- CAS Number:51404-69-4
- Formula:(CH₃COO)₂Pb·2Pb(OH)₂
- MDL Number:MFCD00011154
- Molecular Weight:807.72 g/mol
- Storage Temperature:Ambient
- UN:2291
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Loss on drying:≤1.5 % (105°C, 2 h)
- Titration:≥33.0 % Basic lead (PbO)
- Chloride (Cl):≤0.003 %
- Nitrate (NO3):≤0.003 %
- Calcium (Ca):≤0.01 %
- Copper (Cu):≤0.002 %
- Iron (Fe):≤0.002 %
- Potassium (K):≤0.02 %
- Sodium (Na):≤0.05 %
- Insoluble matter:≤0.02 % (in dilute acetic acid)
- Insoluble matter in water:≤1.0 %
Specifications
- Pack type:Plastic bottle
- Pk:5 kg



