Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Lead(II) acetate basic ACS

Plastic bottle, 1 kg
Catalog # ACRO453220010
Irritant
Health
Environment
Lead(II) acetate basic ACS
Catalog # ACRO453220010
CAS Number:  51404-69-4
Lead(II) acetate basic ACS
Lead(II) acetate basic ACS

Plastic bottle, 1 kg

CAS Number:  51404-69-4
Price
64,80 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:51404-69-4
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Storage Temperature:Ambient
  • UN:2291
  • ADR:6.1,III

 

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1.5 % (105°C, 2 h)
  • Titration:
    ≥33.0 % Basic lead (PbO)
  • Chloride (Cl):
    ≤0.003 %
  • Nitrate (NO3):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.01 %
  • Copper (Cu):
    ≤0.002 %
  • Iron (Fe):
    ≤0.002 %
  • Potassium (K):
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.02 % (in dilute acetic acid)
  • Insoluble matter in water:
    ≤1.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications