Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, Technical Grade

Acetone, Technical Grade

Plastic can, 25 L
Catalog # ACRO444150250
Irritant
Flammable
   Safer Human and Environmental Health
Acetone, Technical Grade
Catalog # ACRO444150250
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone, Technical Grade
Acetone, Technical Grade

Plastic can, 25 L

CAS Number:  67-64-1
Price
181,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • UN:1090
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3580 to 1.3600 (20°C, 589 nm)
  • Density:
    0.790 to 0.792 kg/l
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic can
  • Pk:
    25 L
  • Cat. No.:
    ACRO444150250
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.