Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, Technical Grade

Acetone, Technical Grade

Plastic drum, 5 L
Catalog # ACRO444150050
Irritant
Flammable
   Safer Human and Environmental Health
Acetone, Technical Grade
Catalog # ACRO444150050
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone, Technical Grade
Acetone, Technical Grade

Plastic drum, 5 L

CAS Number:  67-64-1
Price
67,90 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • UN:1090
  • ADR:3,II

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3580 to 1.3600 (20°C, 589 nm)
  • Density:
    0.790 to 0.792 kg/l
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Cat. No.:
    ACRO444150050
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications