Agar, qualité Bactériologie
Plastic drum, 5 kg
Fournisseur: Thermo Fisher Scientific
Agar, qualité Bactériologie
N° de catalogue ACRO443570050
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 9002-18-0
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Melting Pt:90 °C
- Storage Temperature:Température ambiante
Documents
ACRO44357
SDS
Specifications
Specification Test Results
- Appearance:White to cream to light yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (1 g, 105°C)
- Solubility:(1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
- Gel strength:≥700 g/cm² (1.5 %)
- Gel point:32 to 38°C (1.5 %)
- Ash:≤5 %
- pH:6.0 to 7.5 (10 % suspension)
Specifications
- Type de conditionnement:Plastic drum
- Cdt:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



