Agar, Bacteriological Grade
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Agar, Bacteriological Grade
Catalog # ACRO443570010
Supplier: Thermo Fisher Scientific
CAS Number: 9002-18-0
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Melting Pt:90 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to cream to light yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (1 g, 105°C)
- Solubility:(1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
- Gel strength:≥700 g/cm² (1.5 %)
- Gel point:32 to 38°C (1.5 %)
- Ash:≤5 %
- pH:6.0 to 7.5 (10 % suspension)
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



