Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Glass bottle, 5 g
Catalog # ACRO407560050
Toxic
Digitonin
Catalog # ACRO407560050
CAS Number:  11024-24-1
Digitonin
Digitonin

Glass bottle, 5 g

CAS Number:  11024-24-1
Price
1 690,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • Melting Pt:240 °C
  • Storage Temperature:Ambient
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤6 % (1 g, 105°C)
  • Appearance of solution:
    (2.5% in warm EtOH) clear colorless to faint yellow
  • Residue after ignition:
    ≤0.3 %
  • Specific optical rotation:
    -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications