Order Entry
Belgium
Orders LinkContactUsLinkComponent
β-Cyclodextrine 98%

β-Cyclodextrine 98%

Glass bottle, 500 g
Catalog # ACRO406005000
β-Cyclodextrine 98%
Catalog # ACRO406005000
CAS Number:  7585-39-9
β-Cyclodextrine 98%
β-Cyclodextrine 98%

Glass bottle, 500 g

CAS Number:  7585-39-9
Price
276,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Melting Pt:310 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤14 % (105°C, 8 hrs)
  • HPLC:
    ≥97.5 % (ELSD)
  • Specific optical rotation:
    +159° to +165° (20°C, 589 nm) (c=1.5, H2O) (on dry substance)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.