β-Cyclodextrine 98%
Glass bottle, 25 g
Fournisseur: Thermo Fisher Scientific
β-Cyclodextrine 98%
N° de catalogue ACRO406000250
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 7585-39-9
Jump to:
Product Details & Documents
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
- CAS Number:7585-39-9
- Formula:C₄₂H₇₀O₃₅
- MDL Number:MFCD00078139
- Molecular Weight:1134.98 g/mol
- Melting Pt:310 °C
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤14 % (105°C, 8 hrs)
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:+159° to +165° (20°C, 589 nm) (c=1.5, H2O) (on dry substance)
Specifications
- Pack type:Glass bottle
- Pk:25 g



