Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cycloheximide 95%

Cycloheximide 95%

Glass bottle, 25 g
Catalog # ACRO357420250
Toxic
Health
Environment
Cycloheximide 95%
Catalog # ACRO357420250
CAS Number:  66-81-9
Cycloheximide 95%
Cycloheximide 95%

Glass bottle, 25 g

CAS Number:  66-81-9
Price
1 060,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Melting Pt:107…114 °C
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    108 to 118 °C
  • HPLC:
    ≥94 %
  • Specific optical rotation:
    -25° to -32° (20 °C, 589 nm) (c=1, CHCl3)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications