Order Entry
Belgium
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

Fibre drum, 10 kg
Catalog # 349670100.
Health
Celite® 545, filter aid
Catalog # 349670100.
CAS Number:  68855-54-9
Celite® 545, filter aid
Celite® 545, filter aid

Fibre drum, 10 kg

CAS Number:  68855-54-9
Price
278,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • Storage Temperature:Ambient
  • EINECS:272-489-0
Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Particle size:
    ≤25 % (Retained on 150 mesh)
  • Wet density:
    255 to 370 g/l
  • pH:
    9.0 to 11.0 (10 % slurry)
  • Permeability:
    2.6 to 6.5 Darcy
  • Calcium oxide (CaO):
    0.3 % (Typical)
  • Magnesium oxide (MgO):
    0.3 % (Typical)
  • Silicon dioxide (SiO2):
    91.5 % (Typical)
  • Aluminium oxide (Al2O3):
    1.0 % (Typical)
  • Iron(III)oxide (Fe2O3):
    1.5 % (Typical)

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.