Polysorbate 20
Plastic drum, 25 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Boiling Pt:>100 °C (760 mmHg)
- Density:1.1 g/cm³
- Flash Pt:>150 °C
- Storage Temperature:Ambient
- EINECS:500-018-3
Documents
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4720 (20°C, 589 nm)
- Water:≤3 %
- Dioxane:≤10 ppm
- Acid value:≤2 mg KOH/g
- Hydroxyl value:96 to 108 mg KOH/g
- Saponification value:40 to 50 mg KOH/g
Specifications
- Pack type:Plastic drum
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



