Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Chlorophenol 99+%

4-Chlorophenol 99+%

Plastic drum, 5 kg
Catalog # ACRO181000051
Irritant
Environment
4-Chlorophenol 99+%
Catalog # ACRO181000051
CAS Number:  106-48-9
4-Chlorophenol 99+%
4-Chlorophenol 99+%

Plastic drum, 5 kg

CAS Number:  106-48-9
Price
387,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:220 °C (760 mmHg)
  • Melting Pt:41…45 °C
  • Density:1.26 g/cm³
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light brown Crystals or fused mass
  • Infrared spectrum:
    Conforms
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 kg

Specifications