Order Entry
Belgium
Orders LinkContactUsLinkComponent
Creatinine 98%

Creatinine 98%

Glass bottle, 250 g
Catalog # ACRO228942500
Creatinine 98%
Catalog # ACRO228942500
CAS Number:  60-27-5
Creatinine 98%
Creatinine 98%

Glass bottle, 250 g

CAS Number:  60-27-5
Price
271,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Melting Pt:255 °C (dec.)
  • Flash Pt:290 °C
  • Storage Temperature:Ambient
  • EINECS:200-466-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellowish-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Sulfated ash:
    ≤0.2 %
  • Solubility:
    (5 % in water) Clear colorless to slightly yellow
  • Titration with HClO4:
    97.5 to 102.5 % (on dried substance)
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g

Specifications