Quinoline 99%
Glass bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:13,08160
- Boiling Pt:237 °C (760 mmHg)
- Melting Pt:–15 °C
- Density:1.095 g/cm³
- Flash Pt:101 °C
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to dark brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6230 to 1.6270 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg



