D-(-)-Aspartic acid 99+%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
D-(-)-Aspartic acid 99+%
Catalog # ACRO204260050
Supplier: Thermo Fisher Scientific
CAS Number: 1783-96-6
Jump to:
Product Details & Documents
Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- CAS Number:1783-96-6
- Formula:C₄H₇NO₄
- MDL Number:MFCD00063081
- Molecular Weight:133.1 g/mol
- Merck Index:13,00845
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:217-234-6
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.4 % (105°C, 3 hrs)
- Enantiomeric excess:≤0.5 % (L-enantiomer)
- Residue after ignition:≤0.1 %
- Specific optical rotation:-23.8° to -25.8° (20°C, 589 nm) (c=5, 5 N HCl)
- Titration with NaOH:≥99.0 %
- Other amino acids:≤0.3 % (TLC)
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



