Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

2-Phenylethanol 99%

Glass bottle, 250 mL
Catalog # ACRO130182500
Irritant
2-Phenylethanol 99%
Catalog # ACRO130182500
CAS Number:  60-12-8
2-Phenylethanol 99%
2-Phenylethanol 99%

Glass bottle, 250 mL

CAS Number:  60-12-8
Price
36,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Merck Index:12,07370
  • Boiling Pt:219 °C (760 mmHg)
  • Melting Pt:–27 °C
  • Density:1.02 g/cm³
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • EINECS:200-456-2
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5300 to 1.5350 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific gravity:
    (25/25°C) 1.016 to 1.021
  • UV:
    at 220 nm A: ≤0.04
  • Titration after Ion exchange:
    ≥99.0 % (on dry substance)
  • Membrane filtration test:
    (on 0.2 M soln.) Passes test
  • Suitability:
    (HPLC gradient) Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications