Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol 97%

Plastic bottle, 100 g
Catalog # ACRO132731000
D-(-)-Sorbitol 97%
Catalog # ACRO132731000
CAS Number:  50-70-4
D-(-)-Sorbitol 97%
D-(-)-Sorbitol 97%

Plastic bottle, 100 g

CAS Number:  50-70-4
Price
35,70 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:95…99 °C
  • Density:1,524 g/cm³ (20 °C)
  • Flash Pt:≥ 100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1.5 %
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥96.0 % (on anhydrous substance)
  • Specific optical rotation:
    +4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance)
  • Lead (Pb):
    ≤0.5 ppm
  • Nickel (Ni):
    ≤1 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.