2-Phenylethanol 99%
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:219 °C (760 mmHg)
- Melting Pt:–27 °C
- Density:1.02 g/cm³
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:200-456-2
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5300 to 1.5350 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(25/25°C) 1.016 to 1.021
- UV:at 220 nm A: ≤0.04
- Titration after Ion exchange:≥99.0 % (on dry substance)
- Membrane filtration test:(on 0.2 M soln.) Passes test
- Suitability:(HPLC gradient) Passes test
Specifications
- Pack type:Plastic drum
- Pk:10 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



