Pyridine 99+%, extra pure
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure
Catalog # ACRO131780250
Supplier: Thermo Fisher Scientific
CAS Number: 110-86-1
Jump to:
Product Details & Documents
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Merck Index:13,08060
- Boiling Pt:115…116 °C (760 mmHg)
- Melting Pt:–42 °C
- Density:0.978 g/cm³
- Flash Pt:17 °C
- Storage Temperature:Ambient
- EINECS:203-809-9
- UN:1282
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5080 to 1.5110 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤40 APHA
- Residue after evaporation:≤0.02 %
Specifications
- Pack type:Metal drum
- Pk:25 L



