D(+)-Galactose 99+%
Fibre drum, 25 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
- CAS Number:59-23-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00151230
- Molecular Weight:180.16 g/mol
- Merck Index:13,04356
- Boiling Pt:527 °C (1013 hPa)
- Melting Pt:165…168 °C
- Density:1,5 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-416-4
Documents
ACRO15061
SDS
ACRO150610250_TDS_EN
Product specification
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤0.3 % (K.F.)
- Sulfated ash:≤0.1 % (on dry substance)
- Solubility:(10 % in water) Clear colorless
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:+78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)
Specifications
- Type de conditionnement:Fibre drum
- Cdt:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



