Order Entry
Belgium
Orders LinkContactUsLinkComponent
Docusate sodium 96%

Docusate sodium 96%

Plastic drum, 2,5 kg
Catalog # ACRO117100025
Corrosive
Docusate sodium 96%
Catalog # ACRO117100025
CAS Number:  577-11-7
Docusate sodium 96%
Docusate sodium 96%

Plastic drum, 2,5 kg

CAS Number:  577-11-7
Price
229,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • MDL Number:MFCD00012455
  • Molecular Weight:444.57 g/mol
  • Merck Index:12,03460
  • Boiling Pt:? 180 °C (1013 hPa)
  • Melting Pt:153…157 °C
  • Density:~ 1,1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-406-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Waxy solid
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥95 %
  • Water:
    ≤2 %
  • Color of alcohol solution:
    ≤100 APHA (50 % solids in 1:1 alcohol/water)
  • Insoluble matter:
    ≤300 ppm (50 % in toluene)
  • Iodine value:
    ≤0.25 (on solids basis)
  • Acid number:
    ≤2.5 (on solids basis)

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 kg

Specifications